tert-Butyl (4-acetyl-2-fluorophenyl)carbamate 95+%

Lieferant: Apollo Scientific

PC409527-1G PC409527-5G
APOSPC409527-1GEA 429 EUR
APOSPC409527-1G APOSPC409527-5G
tert-Butyl (4-acetyl-2-fluorophenyl)carbamate 95+%
tert-Butyl (4-acetyl-2-fluorophenyl)carbamate
Formel: C₁₃H₁₆FNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD20921817
CAS-Nummer: 811799-54-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR