1-[4-Chloro-3-(trifluoromethyl)phenyl]-2-thiourea 98%

Lieferant: Apollo Scientific

PC4097-10G PC4097-1G PC4097-5G
APOSPC4097-10GEA 495 EUR
APOSPC4097-10G APOSPC4097-1G APOSPC4097-5G
1-[4-Chloro-3-(trifluoromethyl)phenyl]-2-thiourea 98%
1-[4-Chloro-3-(trifluoromethyl)phenyl]-2-thiourea
Formel: C₈H₆ClF₃N₂S
Schmelzpunkt: 139…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041182
CAS-Nummer: 207919-03-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR