1-(2-Fluorobenzyl)-1H-pyrazol-3-ylamine

Lieferant: Apollo Scientific

Synonyme: 1-(2-Fluorobenzyl)-1H-pyrazol-3-amine hydrochloride

PC410001-1G
APOSPC410001-1GEA 232 EUR
APOSPC410001-1G
1-(2-Fluorobenzyl)-1H-pyrazol-3-ylamine
1-(2-Fluorobenzyl)-1H-pyrazol-3-ylamine
Formel: C₁₀H₁₀FN₃
Molecular Weight: 191,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02254004
CAS-Nummer: 895929-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR