1-[3-(Trifluoromethyl)benzyl]-1H-pyrazol-4-amine dihydrochloride

Lieferant: Apollo Scientific

PC410013-1G
APOSPC410013-1GEA 386 EUR
APOSPC410013-1G
1-[3-(Trifluoromethyl)benzyl]-1H-pyrazol-4-amine dihydrochloride
1-[3-(Trifluoromethyl)benzyl]-1H-pyrazol-4-amine dihydrochloride
Formel: C₁₁H₁₀F₃N₃·2ClH
Molecular Weight: 314,134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04967279
CAS-Nummer: 1002033-51-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR