4-(Difluoromethoxy)-3-methoxyaniline hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Difluoromethoxy)-m-anisidine hydrochloride, 5-Amino-2-(difluoromethoxy)anisole hydrochloride

PC410014-1G
APOSPC410014-1GEA 190 EUR
APOSPC410014-1G
4-(Difluoromethoxy)-3-methoxyaniline hydrochloride
4-(Difluoromethoxy)-3-methoxyaniline hydrochloride
Formel: C₈H₉F₂NO₂·ClH
Molecular Weight: 225,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04967289
CAS-Nummer: 1185012-61-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR