1-[(2-Chloro-4-fluorophenoxy)methyl]-4-nitro-1H-pyrazole
Lieferant: Apollo Scientific
PC410024-1G
APOSPC410024-1GEA
386
EUR
APOSPC410024-1G
1-[(2-Chloro-4-fluorophenoxy)methyl]-4-nitro-1H-pyrazole
1-[(2-Chloro-4-fluorophenoxy)methyl]-4-nitro-1H-pyrazole
Formel:
C₁₀H₇ClFN₃O₃ Molecular Weight: 271,632 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06740551 CAS-Nummer: 1003993-40-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...