4-Bromo-1-(4-fluorobenzyl)-3-nitro-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-1-[(4-fluorophenyl)methyl]-3-nitro-1H-pyrazole

PC410027-1G
APOSPC410027-1GEA 516 EUR
APOSPC410027-1G
4-Bromo-1-(4-fluorobenzyl)-3-nitro-1H-pyrazole
4-Bromo-1-(4-fluorobenzyl)-3-nitro-1H-pyrazole
Formel: C₁₀H₇BrFN₃O₂
Molecular Weight: 300.084 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03468600
CAS-Nummer: 512824-95-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR