1-[(2,2,2-Trifluoroethoxy)methyl]-1H-pyrazol-4-amine hydrochloride
Lieferant: Apollo Scientific
PC410032-1G
APOSPC410032-1GEA
232
EUR
APOSPC410032-1G
1-[(2,2,2-Trifluoroethoxy)methyl]-1H-pyrazol-4-amine hydrochloride
1-[(2,2,2-Trifluoroethoxy)methyl]-1H-pyrazol-4-amine hydrochloride
Formel:
C₆H₈F₃N₃O·ClH Molecular Weight: 231,603 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04969858 CAS-Nummer: 1185098-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...