4-[4-(Difluoromethoxy)phenyl]-2-hydrazinylpyrimidine

Lieferant: Apollo Scientific

PC410045-1G
APOSPC410045-1GEA 386 EUR
APOSPC410045-1G
4-[4-(Difluoromethoxy)phenyl]-2-hydrazinylpyrimidine
4-[4-(Difluoromethoxy)phenyl]-2-hydrazinylpyrimidine
Formel: C₁₁H₁₀F₂N₄O
Molecular Weight: 252,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08556206
CAS-Nummer: 932240-78-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR