4-(3-Fluorophenoxy)benzonitrile
Lieferant: Apollo Scientific
PC410057-1G
APOSPC410057-1GEA
386
EUR
APOSPC410057-1G
4-(3-Fluorophenoxy)benzonitrile
4-(3-Fluorophenoxy)benzonitrile
Formel:
C₁₃H₈FNO Molecular Weight: 213,207 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08559040 CAS-Nummer: 877149-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...