4-Chloro-1-[(2,5-difluorophenoxy)methyl]-3-nitro-1H-pyrazole

Lieferant: Apollo Scientific

PC410061-1G
APOSPC410061-1GEA 516 EUR
APOSPC410061-1G
4-Chloro-1-[(2,5-difluorophenoxy)methyl]-3-nitro-1H-pyrazole
4-Chloro-1-[(2,5-difluorophenoxy)methyl]-3-nitro-1H-pyrazole
Formel: C₁₀H₆ClF₂N₃O₃
Molecular Weight: 289,623 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08558152
CAS-Nummer: 1020724-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR