4-Chloro-1-[(2-fluorophenoxy)methyl]-3-nitro-1H-pyrazole
Lieferant: Apollo Scientific
PC410062-1G
APOSPC410062-1GEA
386
EUR
APOSPC410062-1G
4-Chloro-1-[(2-fluorophenoxy)methyl]-3-nitro-1H-pyrazole
4-Chloro-1-[(2-fluorophenoxy)methyl]-3-nitro-1H-pyrazole
Formel:
C₁₀H₇ClFN₃O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08558153 CAS-Nummer: 1020724-26-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...