4-(2,2-Difluoroethoxy)-2-methylaniline hydrochloride
Lieferant: Apollo Scientific
PC410075-1G
APOSPC410075-1GEA
190
EUR
APOSPC410075-1G
4-(2,2-Difluoroethoxy)-2-methylaniline hydrochloride
4-(2,2-Difluoroethoxy)-2-methylaniline hydrochloride
Formel:
C₉H₁₁F₂NO·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD16621777 CAS-Nummer: 1431965-01-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...