4-(2,2,3,3-Tetrafluoropropoxy)aniline hydrochloride

Lieferant: Apollo Scientific

PC410076-1G
APOSPC410076-1GEA 516 EUR
APOSPC410076-1G
4-(2,2,3,3-Tetrafluoropropoxy)aniline hydrochloride
4-(2,2,3,3-Tetrafluoropropoxy)aniline hydrochloride
Formel: C₉H₉F₄NO·ClH
Molecular Weight: 259,628 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25371100
CAS-Nummer: 109230-81-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR