2-(2,2,2-Trifluoroethoxy)aniline hydrochloride

Lieferant: Apollo Scientific

PC410081-1G
APOSPC410081-1GEA 0 EUR
APOSPC410081-1G
2-(2,2,2-Trifluoroethoxy)aniline hydrochloride
2-(2,2,2-Trifluoroethoxy)aniline hydrochloride
Formel: C₈H₈F₃NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD11100362
CAS-Nummer: 62149-83-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR