1-(2,2-Difluoroethyl)-1H-pyrazol-4-amine hydrochloride

Lieferant: Apollo Scientific

PC410084-1G
APOSPC410084-1GEA 386 EUR
APOSPC410084-1G
1-(2,2-Difluoroethyl)-1H-pyrazol-4-amine hydrochloride
1-(2,2-Difluoroethyl)-1H-pyrazol-4-amine hydrochloride
Formel: C₅H₈ClF₂N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD15142911
CAS-Nummer: 1197231-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR