1-[(2-Fluorophenoxy)methyl]-1H-pyrazol-4-amine hydrochloride

Lieferant: Apollo Scientific

PC410085-1G
APOSPC410085-1GEA 647 EUR
APOSPC410085-1G
1-[(2-Fluorophenoxy)methyl]-1H-pyrazol-4-amine hydrochloride
1-[(2-Fluorophenoxy)methyl]-1H-pyrazol-4-amine hydrochloride
Formel: C₁₀H₁₀FN₃O·ClH
Molecular Weight: 243,665 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25370980
CAS-Nummer: 1431963-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR