1-(4-Fluorobenzyl)piperidine-3-carboxylic acid hydrochloride
Lieferant: Apollo Scientific
PC410087-1G
APOSPC410087-1GEA
386
EUR
APOSPC410087-1G
1-(4-Fluorobenzyl)piperidine-3-carboxylic acid hydrochloride
1-(4-Fluorobenzyl)piperidine-3-carboxylic acid hydrochloride
Formel:
C₁₃H₁₆FNO₂·ClH Molecular Weight: 273,731 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06800730 CAS-Nummer: 1185303-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...