1-(2,2-Difluoroethyl)piperazine dihydrochloride

Lieferant: Apollo Scientific

PC410091-1G
APOSPC410091-1GEA 647 EUR
APOSPC410091-1G
1-(2,2-Difluoroethyl)piperazine dihydrochloride
1-(2,2-Difluoroethyl)piperazine dihydrochloride
Formel: C₆H₁₂F₂N₂·2ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD25371148
CAS-Nummer: 1225276-98-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR