3-Fluoro-4-(2,2,2-trifluoroethoxy)aniline hydrochloride

Lieferant: Apollo Scientific

PC410092-1G
APOSPC410092-1GEA 647 EUR
APOSPC410092-1G
3-Fluoro-4-(2,2,2-trifluoroethoxy)aniline hydrochloride
3-Fluoro-4-(2,2,2-trifluoroethoxy)aniline hydrochloride
Formel: C₈H₇F₄NO·ClH
Molecular Weight: 245,602 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25371152
CAS-Nummer: 1431963-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR