1-(2-Chloro-6-fluorobenzyl)-3-nitro-1H-pyrazole
Lieferant: Apollo Scientific
PC410096-1G
APOSPC410096-1GEA
386
EUR
APOSPC410096-1G
1-(2-Chloro-6-fluorobenzyl)-3-nitro-1H-pyrazole
1-(2-Chloro-6-fluorobenzyl)-3-nitro-1H-pyrazole
Formel:
C₁₀H₇ClFN₃O₂ Molecular Weight: 255,633 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03087893 CAS-Nummer: 512823-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...