3-Fluoro-4-(piperidin-1-yl)aniline hydrochloride

Lieferant: Apollo Scientific

PC410098-1G
APOSPC410098-1GEA 232 EUR
APOSPC410098-1G
3-Fluoro-4-(piperidin-1-yl)aniline hydrochloride
3-Fluoro-4-(piperidin-1-yl)aniline hydrochloride
Formel: C₁₁H₁₅FN₂·ClH
Molecular Weight: 230,71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16294066
CAS-Nummer: 1245569-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR