2-Amino-4-(4-fluorophenyl)thiophene-3-carboxamide

Lieferant: Apollo Scientific

PC410102-1G
APOSPC410102-1GEA 516 EUR
APOSPC410102-1G
2-Amino-4-(4-fluorophenyl)thiophene-3-carboxamide
2-Amino-4-(4-fluorophenyl)thiophene-3-carboxamide
Formel: C₁₁H₉FN₂OS
Molecular Weight: 236,265 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02855014
CAS-Nummer: 61019-24-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR