2-(2,2,2-Trifluoroethoxy)pyridin-3-amine hydrochloride

Lieferant: Apollo Scientific

PC410106-1G
APOSPC410106-1GEA 828 EUR
APOSPC410106-1G
2-(2,2,2-Trifluoroethoxy)pyridin-3-amine hydrochloride
2-(2,2,2-Trifluoroethoxy)pyridin-3-amine hydrochloride
Formel: C₇H₇F₃N₂O·ClH
Molecular Weight: 228,599 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25368944
CAS-Nummer: 1431964-68-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR