5-Amino-2-(2,2-difluoroethoxy)pyridine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
PC410107-1G
APOSPC410107-1GEA
828
EUR
APOSPC410107-1G
5-Amino-2-(2,2-difluoroethoxy)pyridine hydrochloride
5-Amino-2-(2,2-difluoroethoxy)pyridine hydrochloride
Formel:
C₇H₈F₂N₂O·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD25371174 CAS-Nummer: 1431962-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...