3-Fluoro-4-[2-(pyrrolidin-1-yl)ethoxy]aniline hydrochloride

Lieferant: Apollo Scientific

PC410109-1G
APOSPC410109-1GEA 232 EUR
APOSPC410109-1G
3-Fluoro-4-[2-(pyrrolidin-1-yl)ethoxy]aniline hydrochloride
3-Fluoro-4-[2-(pyrrolidin-1-yl)ethoxy]aniline hydrochloride
Formel: C₁₂H₁₇FN₂O·ClH
Molecular Weight: 260,736 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15142916
CAS-Nummer: 837421-94-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR