2-[(2-Fluoro-4-nitrophenoxy)methyl]furan

Lieferant: Apollo Scientific

PC410112-1G
APOSPC410112-1GEA 386 EUR
APOSPC410112-1G
2-[(2-Fluoro-4-nitrophenoxy)methyl]furan
2-[(2-Fluoro-4-nitrophenoxy)methyl]furan
Formel: C₁₁H₈FNO₄
Molecular Weight: 237,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10699092
CAS-Nummer: 1160246-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR