3-Fluoro-4-(furan-2-ylmethoxy)aniline hydrochloride
Lieferant: Apollo Scientific
PC410115-1G
APOSPC410115-1GEA
647
EUR
APOSPC410115-1G
3-Fluoro-4-(furan-2-ylmethoxy)aniline hydrochloride
3-Fluoro-4-(furan-2-ylmethoxy)aniline hydrochloride
Formel:
C₁₁H₁₀FNO₂·ClH Molecular Weight: 243,662 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD25371196 CAS-Nummer: 1431966-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...