4-(Cyclohexyloxy)-3-fluoroaniline hydrochloride

Lieferant: Apollo Scientific

PC410116-1G
APOSPC410116-1GEA 516 EUR
APOSPC410116-1G
4-(Cyclohexyloxy)-3-fluoroaniline hydrochloride
4-(Cyclohexyloxy)-3-fluoroaniline hydrochloride
Formel: C₁₂H₁₆FNO·ClH
Molecular Weight: 245,721 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11499050
CAS-Nummer: 1197234-19-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR