4-(3-Methoxyphenyl)-2-(piperazin-1-yl)-6-(trifluoromethyl)pyrimidine dihydrochloride
Lieferant: Apollo Scientific
PC410119-1G
APOSPC410119-1GEA
828
EUR
APOSPC410119-1G
4-(3-Methoxyphenyl)-2-(piperazin-1-yl)-6-(trifluoromethyl)pyrimidine dihydrochloride
4-(3-Methoxyphenyl)-2-(piperazin-1-yl)-6-(trifluoromethyl)pyrimidine dihydrochloride
Formel:
C₁₆H₁₇F₃N₄O·2ClH Molecular Weight: 411,249 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD25371203 CAS-Nummer: 1431970-11-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...