5-Fluoro-1H-indole-3-carbonitrile 98%

Lieferant: Apollo Scientific
Achtung

PC410190-1G
APOSPC410190-1GEA 145 EUR
APOSPC410190-1G
5-Fluoro-1H-indole-3-carbonitrile 98%
5-Fluoro-1H-indole-3-carbonitrile
Formel: C9H5FN2
Molecular Weight: 160,1500 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06809706
CAS-Nummer: 194490-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR