4-Chloro-1-(2-fluorobenzyl)-3-nitro-1H-pyrazole
Lieferant: Apollo Scientific
PC410259-1G
APOSPC410259-1GEA
386
EUR
APOSPC410259-1G
4-Chloro-1-(2-fluorobenzyl)-3-nitro-1H-pyrazole
4-Chloro-1-(2-fluorobenzyl)-3-nitro-1H-pyrazole
Formel:
C₁₀H₇ClFN₃O₂ Molecular Weight: 255,633 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04969178 CAS-Nummer: 1001500-06-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...