[4-Bromo-5-cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetic acid

Lieferant: Apollo Scientific

PC410260-1G
APOSPC410260-1GEA 647 EUR
APOSPC410260-1G
[4-Bromo-5-cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetic acid
[4-Bromo-5-cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetic acid
Formel: C₉H₉BrF₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04969184
CAS-Nummer: 1001500-11-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR