3-Nitro-1-[3-(trifluoromethyl)benzyl]-1H-pyrazole
Lieferant: Apollo Scientific
PC410263-1G
APOSPC410263-1GEA
0
EUR
APOSPC410263-1G
3-Nitro-1-[3-(trifluoromethyl)benzyl]-1H-pyrazole
3-Nitro-1-[3-(trifluoromethyl)benzyl]-1H-pyrazole
Formel:
C₁₁H₈F₃N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD02029256 CAS-Nummer: 1001510-37-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...