4-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-5-carbohydrazide

Lieferant: Apollo Scientific

PC410265-1G
APOSPC410265-1GEA 516 EUR
APOSPC410265-1G
4-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-5-carbohydrazide
4-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-5-carbohydrazide
Formel: C₆H₆ClF₃N₄O
Molecular Weight: 242.586 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419675
CAS-Nummer: 1001518-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR