4-Bromo-1-methyl-3-(trifluoromethyl)-1H-pyrazole-5-carbohydrazide

Lieferant: Apollo Scientific

PC410266-1G
APOSPC410266-1GEA 386 EUR
APOSPC410266-1G
4-Bromo-1-methyl-3-(trifluoromethyl)-1H-pyrazole-5-carbohydrazide
4-Bromo-1-methyl-3-(trifluoromethyl)-1H-pyrazole-5-carbohydrazide
Formel: C₆H₆BrF₃N₄O
Molecular Weight: 287,037 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419676
CAS-Nummer: 1001518-80-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR