[4-Bromo-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]acetic acid

Lieferant: Apollo Scientific

PC410267-1G
APOSPC410267-1GEA 647 EUR
APOSPC410267-1G
[4-Bromo-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]acetic acid
[4-Bromo-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]acetic acid
Formel: C₇H₇BrF₂N₂O₂
Molecular Weight: 269.043 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419678
CAS-Nummer: 1001518-81-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR