2-[3-(Difluoromethyl)-5-methyl-1H-pyrazol-1-yl]acetohydrazide
Lieferant: Apollo Scientific
PC410268-1G
APOSPC410268-1GEA
386
EUR
APOSPC410268-1G
2-[3-(Difluoromethyl)-5-methyl-1H-pyrazol-1-yl]acetohydrazide
2-[3-(Difluoromethyl)-5-methyl-1H-pyrazol-1-yl]acetohydrazide
Formel:
C₇H₁₀F₂N₄O Molecular Weight: 204.177 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03419679 CAS-Nummer: 1001518-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...