2-[4-Bromo-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]acetohydrazide
Lieferant: Apollo Scientific
PC410269-1G
APOSPC410269-1GEA
0
EUR
APOSPC410269-1G
2-[4-Bromo-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]acetohydrazide
2-[4-Bromo-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]acetohydrazide
Formel:
C₇H₉BrF₂N₄O Lagertemperatur: Raumtemperatur |
MDL:
MFCD03419681 CAS-Nummer: 1001518-83-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...