3-[4-Chloro-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid

Lieferant: Apollo Scientific
Achtung

PC410270-1G
APOSPC410270-1GEA 647 EUR
APOSPC410270-1G
3-[4-Chloro-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
3-[4-Chloro-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Formel: C₈H₈ClF₃N₂O₂
Molecular Weight: 256.61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419694
CAS-Nummer: 1001518-84-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR