3-[4-Bromo-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid

Lieferant: Apollo Scientific

PC410271-1G
APOSPC410271-1GEA 516 EUR
APOSPC410271-1G
3-[4-Bromo-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
3-[4-Bromo-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Formel: C₈H₈BrF₃N₂O₂
Molecular Weight: 301,061 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419695
CAS-Nummer: 1001518-85-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR