3-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide

Lieferant: Apollo Scientific

PC410272-1G
APOSPC410272-1GEA 232 EUR
APOSPC410272-1G
3-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
3-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
Formel: C₈H₁₁F₃N₄O
Molecular Weight: 236,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419696
CAS-Nummer: 1001518-86-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR