2-[3,5-Bis(difluoromethyl)-1H-pyrazol-1-yl]acetohydrazide

Lieferant: Apollo Scientific

PC410273-1G
APOSPC410273-1GEA 516 EUR
APOSPC410273-1G
2-[3,5-Bis(difluoromethyl)-1H-pyrazol-1-yl]acetohydrazide
2-[3,5-Bis(difluoromethyl)-1H-pyrazol-1-yl]acetohydrazide
Formel: C₇H₈F₄N₄O
Molecular Weight: 240.158 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419713
CAS-Nummer: 1001518-87-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR