2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetohydrazide

Lieferant: Apollo Scientific

PC410279-1G
APOSPC410279-1GEA 232 EUR
APOSPC410279-1G
2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetohydrazide
2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetohydrazide
Formel: C₉H₁₁F₃N₄O
Molecular Weight: 248,205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419742
CAS-Nummer: 1001518-93-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR