2-[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetohydrazide

Lieferant: Apollo Scientific

PC410280-1G
APOSPC410280-1GEA 386 EUR
APOSPC410280-1G
2-[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetohydrazide
2-[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetohydrazide
Formel: C₉H₁₀ClF₃N₄O
Molecular Weight: 282,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419743
CAS-Nummer: 1001518-94-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR