3-[4-Bromo-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid

Lieferant: Apollo Scientific

PC410282-1G
APOSPC410282-1GEA 516 EUR
APOSPC410282-1G
3-[4-Bromo-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
3-[4-Bromo-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Formel: C₁₀H₁₀BrF₃N₂O₂
Molecular Weight: 327.098 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419753
CAS-Nummer: 1001518-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR