3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide

Lieferant: Apollo Scientific

PC410283-1G
APOSPC410283-1GEA 386 EUR
APOSPC410283-1G
3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
Formel: C₁₀H₁₃F₃N₄O
Molecular Weight: 262.232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419754
CAS-Nummer: 1001518-97-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR