3-[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide

Lieferant: Apollo Scientific

PC410284-1G
APOSPC410284-1GEA 0 EUR
APOSPC410284-1G
3-[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
3-[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
Formel: C₁₀H₁₂ClF₃N₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD03419755
CAS-Nummer: 1001518-98-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR