2-Methoxy-4-(1-methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidine

Lieferant: Apollo Scientific

PC410285-1G
APOSPC410285-1GEA 386 EUR
APOSPC410285-1G
2-Methoxy-4-(1-methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidine
2-Methoxy-4-(1-methyl-1H-pyrazol-4-yl)-6-(trifluoromethyl)pyrimidine
Formel: C₁₀H₉F₃N₄O
Molecular Weight: 258,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05667172
CAS-Nummer: 1001519-12-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR