1-Ethyl-5-(trifluoromethyl)-1H-pyrazole-3-carbohydrazide

Lieferant: Apollo Scientific

PC410286-1G
APOSPC410286-1GEA 516 EUR
APOSPC410286-1G
1-Ethyl-5-(trifluoromethyl)-1H-pyrazole-3-carbohydrazide
1-Ethyl-5-(trifluoromethyl)-1H-pyrazole-3-carbohydrazide
Formel: C₇H₉F₃N₄O
Molecular Weight: 222,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05667186
CAS-Nummer: 1001519-13-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR